Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGAAAACCTCACCGAGCAGCCT[A/G]AGCAGCTGTCACCACCTTCATCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDFY3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDFY3 - WD repeat and FYVE domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014991.4 | 10742 | Missense Mutation | TCA,TTA | S,L 3461 | NP_055806.2 | |
XM_005262858.4 | 10742 | Missense Mutation | TCA,TTA | S,L 3479 | XP_005262915.1 | |
XM_011531757.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3479 | XP_011530059.1 | |
XM_011531759.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3479 | XP_011530061.1 | |
XM_011531760.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3479 | XP_011530062.1 | |
XM_011531761.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3464 | XP_011530063.1 | |
XM_011531762.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3462 | XP_011530064.1 | |
XM_011531763.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3461 | XP_011530065.1 | |
XM_011531764.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3451 | XP_011530066.1 | |
XM_011531765.2 | 10742 | Missense Mutation | TCA,TTA | S,L 3444 | XP_011530067.1 | |
XM_011531766.2 | 10742 | Intron | XP_011530068.1 | |||
XM_011531767.2 | 10742 | Intron | XP_011530069.1 | |||
XM_017007906.1 | 10742 | Missense Mutation | TCA,TTA | S,L 3479 | XP_016863395.1 | |
XM_017007907.1 | 10742 | Missense Mutation | TCA,TTA | S,L 3479 | XP_016863396.1 | |
XM_017007908.1 | 10742 | Missense Mutation | TCA,TTA | S,L 3479 | XP_016863397.1 | |
XM_017007909.1 | 10742 | Missense Mutation | TCA,TTA | S,L 2801 | XP_016863398.1 |