Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGTCTCTCTCTCTGGCCTTGGC[A/C]TCTGTGACTATCAGGTCCTCGCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606022 | ||||||||||||||||||||
Literature Links: |
NUDT9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUDT9 - nudix hydrolase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248011.1 | 393 | Silent Mutation | GCA,GCC | A,A 20 | NP_001234940.1 | |
NM_024047.4 | 393 | Silent Mutation | GCA,GCC | A,A 20 | NP_076952.1 | |
NM_198038.2 | 393 | Intron | NP_932155.1 | |||
XM_011532031.2 | 393 | Silent Mutation | GCA,GCC | A,A 20 | XP_011530333.1 | |
XM_011532032.2 | 393 | Intron | XP_011530334.1 |