Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCTGTAAATCTTCTACCAGAGT[C/T]TGCAAGCTTTCTAGTCTGTTCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC158 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC158 - coiled-coil domain containing 158 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042784.1 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1077 | NP_001036249.1 | |
XM_005262974.4 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1077 | XP_005263031.1 | |
XM_006714204.3 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_006714267.1 | |
XM_011531908.2 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_011530210.1 | |
XM_011531909.2 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_011530211.1 | |
XM_011531910.2 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_011530212.1 | |
XM_011531911.1 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_011530213.1 | |
XM_011531912.2 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_011530214.1 | |
XM_011531913.1 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_011530215.1 | |
XM_011531914.2 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_011530216.1 | |
XM_011531915.2 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1030 | XP_011530217.1 | |
XM_011531916.2 | 3384 | Silent Mutation | CAA,CAG | Q,Q 999 | XP_011530218.1 | |
XM_011531917.1 | 3384 | Silent Mutation | CAA,CAG | Q,Q 999 | XP_011530219.1 | |
XM_017008160.1 | 3384 | Silent Mutation | CAA,CAG | Q,Q 1081 | XP_016863649.1 | |
XM_017008161.1 | 3384 | Silent Mutation | CAA,CAG | Q,Q 781 | XP_016863650.1 |