Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCACCGGGGTGGCCTTGAGGAG[C/T]GTGACTCGGAAGTAGGAACCCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601207 MIM: 616660 | ||||||||||||||||||||
Literature Links: |
DGKQ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGKQ - diacylglycerol kinase theta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001347.3 | 2703 | Silent Mutation | ACA,ACG | T,T 880 | NP_001338.2 | |
XM_011513411.1 | 2703 | Silent Mutation | ACA,ACG | T,T 880 | XP_011511713.1 | |
XM_011513412.1 | 2703 | Intron | XP_011511714.1 | |||
XM_011513414.2 | 2703 | Intron | XP_011511716.1 | |||
XM_011513415.1 | 2703 | Intron | XP_011511717.1 | |||
XM_017007814.1 | 2703 | Intron | XP_016863303.1 | |||
XM_017007815.1 | 2703 | Intron | XP_016863304.1 |
TMEM175 - transmembrane protein 175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |