Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAAGTCACCAGCCTTGGATGGTT[A/G]TCTAACAGGTCAGGCTTAAAGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602598 MIM: 612761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPGDS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPGDS - hematopoietic prostaglandin D synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014485.2 | 598 | Silent Mutation | GAC,GAT | D,D 169 | NP_055300.1 | |
XM_005262932.2 | 598 | Silent Mutation | GAC,GAT | D,D 138 | XP_005262989.1 |
SMARCAD1 - SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |