Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAATCTGAAACATGAGGAGTAC[A/G]ATTCTACTGTTTTGTCTTCTAGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610912 | ||||||||||||||||||||
Literature Links: |
AMTN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMTN - amelotin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286731.1 | 110 | Silent Mutation | ACA,ACG | T,T 4 | NP_001273660.1 | |
NM_212557.3 | 110 | Silent Mutation | ACA,ACG | T,T 4 | NP_997722.1 |