Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGCGGTATAATCAATTCAACTT[C/T]ATGAACGGCCCACATGTAAGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606585 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENAM - enamelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031889.2 | 2225 | Silent Mutation | TTC,TTT | F,F 65 | NP_114095.2 | |
XM_006714056.3 | 2225 | Silent Mutation | TTC,TTT | F,F 65 | XP_006714119.1 |