Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTTACATAGATCCACAAGAAGA[C/T]AGTGGTGCCCAAGGTGAACCCAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608000 MIM: 603844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAA15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAA15 - N(alpha)-acetyltransferase 15, NatA auxiliary subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFC1 - NADH:ubiquinone oxidoreductase subunit C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184986.1 | 587 | Missense Mutation | ATC,GTC | I,V 52 | NP_001171915.1 | |
NM_001184987.1 | 587 | Missense Mutation | ATC,GTC | I,V 52 | NP_001171916.1 | |
NM_001184988.1 | 587 | Missense Mutation | ATC,GTC | I,V 52 | NP_001171917.1 | |
NM_001184989.1 | 587 | Missense Mutation | ATC,GTC | I,V 52 | NP_001171918.1 | |
NM_001184990.1 | 587 | Missense Mutation | ATC,GTC | I,V 52 | NP_001171919.1 | |
NM_001184991.1 | 587 | Missense Mutation | ATC,GTC | I,V 52 | NP_001171920.1 | |
NM_002494.3 | 587 | Missense Mutation | ATC,GTC | I,V 52 | NP_002485.1 |