Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCACAAGAAGAAGATGAGGACT[A/G]TGATTTTGAAGATCGGGTCAGCGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610621 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INTU - inturned planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015693.3 | 216 | Missense Mutation | TAT,TGT | Y,C 29 | NP_056508.2 | |
XM_011531844.2 | 216 | Missense Mutation | TAT,TGT | Y,C 29 | XP_011530146.1 | |
XM_011531845.2 | 216 | Missense Mutation | TAT,TGT | Y,C 29 | XP_011530147.1 | |
XM_011531846.1 | 216 | Intron | XP_011530148.1 | |||
XM_011531847.1 | 216 | Intron | XP_011530149.1 | |||
XM_011531848.2 | 216 | Intron | XP_011530150.1 | |||
XM_011531849.2 | 216 | Missense Mutation | TAT,TGT | Y,C 29 | XP_011530151.1 | |
XM_011531850.2 | 216 | Missense Mutation | TAT,TGT | Y,C 29 | XP_011530152.1 | |
XM_011531851.2 | 216 | Missense Mutation | TAT,TGT | Y,C 29 | XP_011530153.1 | |
XM_017008025.1 | 216 | Intron | XP_016863514.1 | |||
XM_017008026.1 | 216 | Missense Mutation | TAT,TGT | Y,C 29 | XP_016863515.1 |