Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAAGAAGGCAGAAAAGCGGCTG[G/T]TTGACGCCTCATCCTTCGGGAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610796 | ||||||||||||||||||||
Literature Links: |
SLC25A31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A31 - solute carrier family 25 member 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318467.1 | 211 | Missense Mutation | GTT,TTT | V,F 15 | NP_001305396.1 | |
NM_031291.3 | 211 | Missense Mutation | GTT,TTT | V,F 15 | NP_112581.1 | |
XM_011532298.1 | 211 | Missense Mutation | GTT,TTT | V,F 15 | XP_011530600.1 |