Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTTTTCTGCATTTGTAGGGGC[A/C]CGATACAACTGCAGCTGCAATAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608614 | ||||||||||||||||||||
Literature Links: |
CYP4V2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP4V2 - cytochrome P450 family 4 subfamily V member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207352.3 | 1081 | Missense Mutation | CAC,CCC | H,P 331 | NP_997235.3 | |
XM_005262935.3 | 1081 | Missense Mutation | CAC,CCC | H,P 331 | XP_005262992.1 | |
XM_017008037.1 | 1081 | Missense Mutation | CAC,CCC | H,P 199 | XP_016863526.1 |