Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAATATTGCTGCTATTACTGCCA[C/T]TGCTGTCCGTGCCGTTAGTCTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114105 | ||||||||||||||||||||
Literature Links: |
PPP3CA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP3CA - protein phosphatase 3 catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000944.4 | 2057 | Missense Mutation | AAT,AGT | N,S 513 | NP_000935.1 | |
NM_001130691.1 | 2057 | Missense Mutation | AAT,AGT | N,S 503 | NP_001124163.1 | |
NM_001130692.1 | 2057 | Missense Mutation | AAT,AGT | N,S 461 | NP_001124164.1 | |
XM_017008365.1 | 2057 | Missense Mutation | AAT,AGT | N,S 501 | XP_016863854.1 |