Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTACCAGGAACTGTCGGAAAACC[A/G]TTTCTGGTTGTTCTCGCTGCTTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607186 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC24D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC24D - SEC24 homolog D, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318066.1 | 2448 | Missense Mutation | ACG,ATG | T,M 999 | NP_001304995.1 | |
NM_014822.3 | 2448 | Missense Mutation | ACG,ATG | T,M 998 | NP_055637.2 | |
XM_005263379.2 | 2448 | Missense Mutation | ACG,ATG | T,M 999 | XP_005263436.1 | |
XM_017008875.1 | 2448 | Missense Mutation | ACG,ATG | T,M 554 | XP_016864364.1 |