Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTGGAGGCATTCCTGGAGGGAG[G/T]AGCTCCCTGGGGTTTTACTCTAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604570 | ||||||||||||||||||||
Literature Links: |
SHROOM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHROOM3 - shroom family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020859.3 | 855 | Missense Mutation | GGA,GTA | G,V 35 | NP_065910.3 | |
XM_005263163.4 | 855 | Intron | XP_005263220.1 | |||
XM_005263164.4 | 855 | Intron | XP_005263221.1 | |||
XM_005263165.4 | 855 | Intron | XP_005263222.1 | |||
XM_006714282.3 | 855 | Intron | XP_006714345.1 | |||
XM_011532157.2 | 855 | Intron | XP_011530459.2 | |||
XM_011532158.2 | 855 | Missense Mutation | GGA,GTA | G,V 35 | XP_011530460.1 | |
XM_011532159.1 | 855 | Missense Mutation | GGA,GTA | G,V 35 | XP_011530461.1 | |
XM_011532160.1 | 855 | Intron | XP_011530462.1 | |||
XM_011532161.2 | 855 | Intron | XP_011530463.1 | |||
XM_017008488.1 | 855 | Intron | XP_016863977.1 | |||
XM_017008489.1 | 855 | Intron | XP_016863978.1 |