Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCCTCAGGAGCTGGGAGGAACA[C/T]AACACACCTTTATCTTGTCCTGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRIML1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIML1 - tripartite motif family like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178556.3 | 256 | Silent Mutation | CAC,CAT | H,H 47 | NP_848651.2 | |
XM_006714207.3 | 256 | Silent Mutation | CAC,CAT | H,H 47 | XP_006714270.1 | |
XM_011531919.1 | 256 | UTR 5 | XP_011530221.1 |