Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAAGGAAACGTGTGAAGTACTA[C/T]GTGTCCAAAATCGTCATCCATCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS48 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRSS48 - protease, serine 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183375.2 | 740 | Silent Mutation | TAC,TAT | Y,Y 98 | NP_899231.2 | |
XM_011531921.2 | 740 | Silent Mutation | TAC,TAT | Y,Y 121 | XP_011530223.1 | |
XM_011531922.2 | 740 | Intron | XP_011530224.1 |