Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATGGGAGGGCTGATTTTCGAG[A/G]GCCGAATGCAGAGGTTCCAAGACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OCIAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OCIAD1 - OCIA domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079839.2 | 444 | Missense Mutation | GAG,GGG | E,G 9 | NP_001073308.1 | |
NM_001079840.2 | 444 | Missense Mutation | GAG,GGG | E,G 9 | NP_001073309.1 | |
NM_001079841.2 | 444 | Missense Mutation | GAG,GGG | E,G 9 | NP_001073310.1 | |
NM_001079842.2 | 444 | Missense Mutation | GAG,GGG | E,G 9 | NP_001073311.2 | |
NM_001168254.1 | 444 | Missense Mutation | GAG,GGG | E,G 14 | NP_001161726.1 | |
NM_017830.3 | 444 | Missense Mutation | GAG,GGG | E,G 9 | NP_060300.1 | |
XM_017008329.1 | 444 | Missense Mutation | GAG,GGG | E,G 9 | XP_016863818.1 |