Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTCGGGGATGGGGACCTGAATGG[C/T]AGCAGCAGGGAAGGCCTGAGGTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RBM47 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM47 - RNA binding motif protein 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098634.1 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | NP_001092104.1 | |
NM_019027.3 | 2572 | Missense Mutation | ACC,GCC | T,A 512 | NP_061900.2 | |
XM_005248103.3 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_005248160.1 | |
XM_005248107.3 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_005248164.1 | |
XM_005248108.3 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_005248165.1 | |
XM_005248109.4 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_005248166.1 | |
XM_011513704.2 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_011512006.1 | |
XM_011513707.2 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_011512009.1 | |
XM_011513708.2 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_011512010.1 | |
XM_017008304.1 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_016863793.1 | |
XM_017008305.1 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_016863794.1 | |
XM_017008306.1 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_016863795.1 | |
XM_017008307.1 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_016863796.1 | |
XM_017008308.1 | 2572 | Missense Mutation | ACC,GCC | T,A 581 | XP_016863797.1 | |
XM_017008309.1 | 2572 | Missense Mutation | ACC,GCC | T,A 580 | XP_016863798.1 | |
XM_017008310.1 | 2572 | Missense Mutation | ACC,GCC | T,A 512 | XP_016863799.1 |