Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTCTGATTTATGGCTTTCAGAT[A/C]TTTTCTGTTACCAACAACACAGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606178 | ||||||||||||||||||||
Literature Links: |
HHIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HHIP - hedgehog interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022475.2 | 831 | Silent Mutation | ATA,ATC | I,I 94 | NP_071920.1 | |
XM_005263178.4 | 831 | Silent Mutation | ATA,ATC | I,I 94 | XP_005263235.1 | |
XM_006714288.3 | 831 | Silent Mutation | ATA,ATC | I,I 94 | XP_006714351.1 |
HHIP-AS1 - HHIP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |