Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTACCTCATTTCACCAATCAGA[C/T]ATGGAAGATGATGAGGAAGAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF16 - DDB1 and CUL4 associated factor 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAPG - non-SMC condensin I complex subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022346.4 | 514 | Silent Mutation | GAC,GAT | D,D 83 | NP_071741.2 | |
XM_011513876.2 | 514 | UTR 5 | XP_011512178.1 | |||
XM_017008543.1 | 514 | Silent Mutation | GAC,GAT | D,D 83 | XP_016864032.1 | |
XM_017008544.1 | 514 | UTR 5 | XP_016864033.1 |