Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAGGGCAGCGCAGGGTGCCGG[A/G]GAAGCTCGGGAATGGAGGTGGTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611792 | ||||||||||||||||||||
Literature Links: |
ZCCHC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZCCHC4 - zinc finger CCHC-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318148.1 | 95 | Missense Mutation | AGA,GGA | R,G 21 | NP_001305077.1 | |
NM_024936.2 | 95 | Missense Mutation | AGA,GGA | R,G 21 | NP_079212.2 | |
XM_011513835.1 | 95 | Missense Mutation | AGA,GGA | R,G 21 | XP_011512137.1 | |
XM_017008128.1 | 95 | Silent Mutation | GGA,GGG | G,G 15 | XP_016863617.1 | |
XM_017008129.1 | 95 | Missense Mutation | AGA,GGA | R,G 21 | XP_016863618.1 |