Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCAGCATTGAGGTAAGCACAAGG[G/T]TAATTATTTGTATGTGACATAAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613009 | ||||||||||||||||||||
Literature Links: |
SEPSECS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEPSECS - Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016955.3 | 1474 | Nonsense Mutation | NP_058651.3 | |||
XM_011513846.2 | 1474 | Nonsense Mutation | XP_011512148.1 | |||
XM_011513847.2 | 1474 | Nonsense Mutation | XP_011512149.1 | |||
XM_011513848.1 | 1474 | Nonsense Mutation | XP_011512150.1 | |||
XM_017008277.1 | 1474 | Nonsense Mutation | XP_016863766.1 | |||
XM_017008278.1 | 1474 | Nonsense Mutation | XP_016863767.1 |