Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTATAAAATATTAACCTGGTCT[C/T]TGGTGAAACTCAGGGGGATAGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YTHDC1 - YTH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005265706.2 | 2164 | Silent Mutation | CAA,CAG | Q,Q 559 | XP_005265763.1 | |
XM_005265707.2 | 2164 | Silent Mutation | CAA,CAG | Q,Q 544 | XP_005265764.1 | |
XM_005265708.2 | 2164 | Silent Mutation | CAA,CAG | Q,Q 541 | XP_005265765.1 | |
XM_017008832.1 | 2164 | Silent Mutation | CAA,CAG | Q,Q 544 | XP_016864321.1 | |
XM_017008833.1 | 2164 | Silent Mutation | CAA,CAG | Q,Q 526 | XP_016864322.1 |