Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGCTGACCATGTGGACCAAGGA[C/T]GGCCGCACCATCCACAGCGGCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605830 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFRL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGFRL1 - fibroblast growth factor receptor-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004356.2 | 351 | Silent Mutation | GAC,GAT | D,D 66 | NP_001004356.1 | |
NM_001004358.1 | 351 | Silent Mutation | GAC,GAT | D,D 66 | NP_001004358.1 | |
NM_021923.3 | 351 | Silent Mutation | GAC,GAT | D,D 66 | NP_068742.2 | |
XM_011513486.1 | 351 | Silent Mutation | GAC,GAT | D,D 66 | XP_011511788.1 |