Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCCCGTATATAATGGCATTGAG[A/G]CAGCTGTTGAAATACGCCATGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTNR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTNR1A - melatonin receptor 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005958.4 | 1069 | Silent Mutation | TGC,TGT | C,C 289 | NP_005949.1 | |
XM_011532002.2 | 1069 | Silent Mutation | TGC,TGT | C,C 204 | XP_011530304.1 |