Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATTTGAACTTCATCTTCTGCTT[G/T]AAGTTCATGGCATGTACCACTGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612486 | ||||||||||||||||||||
Literature Links: |
DCHS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCHS2 - dachsous cadherin-related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142552.1 | 6019 | Intron | NP_001136024.1 | |||
NM_017639.3 | 6019 | Missense Mutation | AAA,CAA | K,Q 2909 | NP_060109.2 | |
XM_011532045.1 | 6019 | Missense Mutation | AAA,CAA | K,Q 3364 | XP_011530347.1 | |
XM_011532046.2 | 6019 | Missense Mutation | AAA,CAA | K,Q 2587 | XP_011530348.1 | |
XM_011532049.2 | 6019 | Missense Mutation | AAA,CAA | K,Q 1699 | XP_011530351.1 | |
XM_017008320.1 | 6019 | Intron | XP_016863809.1 |