Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAACACAAAGACCAGGCTACAGAG[A/G]CAAGAGGTGTCTGGTGAGAGGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611434 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLNK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLNK - cytokine dependent hematopoietic cell linker | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052964.2 | 1647 | Missense Mutation | CCT,TCT | P,S 427 | NP_443196.2 | |
XM_011513775.2 | 1647 | Missense Mutation | CCT,TCT | P,S 442 | XP_011512077.1 | |
XM_017007684.1 | 1647 | Intron | XP_016863173.1 | |||
XM_017007685.1 | 1647 | Intron | XP_016863174.1 |