Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTATGCACATACCTGGTCTCATCC[A/G]AACCCTGCGGATATATTTTTCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611135 MIM: 607031 MIM: 603686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLB - klotho beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIAS - lipoic acid synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL9 - ribosomal protein L9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000661.4 | Intron | NP_000652.2 | ||||
NM_001024921.3 | Intron | NP_001020092.1 |