Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGCATTGAGTAAGTCTGTACT[C/T]GACATTGTGTGGAAGTCCAAACTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600067 | ||||||||||||||||||||
Literature Links: |
UGT2B4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UGT2B4 - UDP glucuronosyltransferase family 2 member B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297615.1 | 1310 | Intron | NP_001284544.1 | |||
NM_001297616.1 | 1310 | Silent Mutation | NP_001284545.1 | |||
NM_021139.2 | 1310 | Silent Mutation | NP_066962.2 |