Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGACGATGGCCTCTGTGGCTTC[C/G]TGCGATTCGCGTCCGAGCTCAGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610621 | ||||||||||||||||||||
Literature Links: |
INTU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INTU - inturned planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015693.3 | 148 | Silent Mutation | TCC,TCG | S,S 6 | NP_056508.2 | |
XM_011531844.2 | 148 | Silent Mutation | TCC,TCG | S,S 6 | XP_011530146.1 | |
XM_011531845.2 | 148 | Silent Mutation | TCC,TCG | S,S 6 | XP_011530147.1 | |
XM_011531846.1 | 148 | Intron | XP_011530148.1 | |||
XM_011531847.1 | 148 | Intron | XP_011530149.1 | |||
XM_011531848.2 | 148 | Intron | XP_011530150.1 | |||
XM_011531849.2 | 148 | Silent Mutation | TCC,TCG | S,S 6 | XP_011530151.1 | |
XM_011531850.2 | 148 | Silent Mutation | TCC,TCG | S,S 6 | XP_011530152.1 | |
XM_011531851.2 | 148 | Silent Mutation | TCC,TCG | S,S 6 | XP_011530153.1 | |
XM_017008025.1 | 148 | Intron | XP_016863514.1 | |||
XM_017008026.1 | 148 | Silent Mutation | TCC,TCG | S,S 6 | XP_016863515.1 |