Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGTCAGAGGCATTTTCCAGAT[C/G]AGTCTTGTACATGCCAGAGTCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LRRC66 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC66 - leucine rich repeat containing 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024611.2 | 4099 | Missense Mutation | CAT,GAT | H,D 789 | NP_001019782.1 | |
XM_011534383.2 | 4099 | Missense Mutation | CAT,GAT | H,D 841 | XP_011532685.1 | |
XM_017008162.1 | 4099 | Missense Mutation | CAT,GAT | H,D 553 | XP_016863651.1 |