Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTATACAGTACTGGCCTTGCAAT[A/G]GTGGTAAGCTTCTTATTATCTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF330 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF330 - zinc finger protein 330 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292002.1 | 445 | Missense Mutation | ATA,ATG | I,M 36 | NP_001278931.1 | |
NM_014487.5 | 445 | Missense Mutation | ATA,ATG | I,M 96 | NP_055302.1 | |
XM_017008033.1 | 445 | Missense Mutation | ATA,ATG | I,M 96 | XP_016863522.1 |