Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTTGCATGCCCCCAGAGACTTA[A/G]TCTTGCTGCTGATGAATATCCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600421 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLRA3 - glycine receptor alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042543.2 | 1896 | Silent Mutation | GAC,GAT | D,D 449 | NP_001036008.1 | |
NM_006529.3 | 1896 | Silent Mutation | GAC,GAT | D,D 464 | NP_006520.2 | |
XM_011532267.2 | 1896 | Intron | XP_011530569.1 | |||
XM_011532268.2 | 1896 | Silent Mutation | GAC,GAT | D,D 269 | XP_011530570.1 | |
XM_017008630.1 | 1896 | Silent Mutation | GAC,GAT | D,D 424 | XP_016864119.1 |