Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCCGCTCCAACACCTTGCTTTC[C/G]TGCAGGACAGCGCCCCAGAGGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603402 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOX3 - acyl-CoA oxidase 3, pristanoyl | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101667.1 | 2040 | UTR 3 | NP_001095137.1 | |||
NM_003501.2 | 2040 | Silent Mutation | CAC,CAG | H,Q 672 | NP_003492.2 | |
XM_005248011.4 | 2040 | Missense Mutation | CAC,CAG | H,Q 672 | XP_005248068.1 | |
XM_005248013.3 | 2040 | UTR 3 | XP_005248070.1 | |||
XM_011513565.2 | 2040 | Missense Mutation | CAC,CAG | H,Q 672 | XP_011511867.1 | |
XM_011513566.1 | 2040 | Missense Mutation | CAC,CAG | H,Q 591 | XP_011511868.1 |