Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTGGGTATTTTGTTTGGTTACC[C/T]GTTGTCTTCGTTTTTCTTCTTCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC101927179 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927179 - uncharacterized LOC101927179 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFXL1 - nuclear transcription factor, X-box binding like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278623.1 | 2658 | Missense Mutation | CAG,CGG | Q,R 854 | NP_001265552.1 | |
NM_001278624.1 | 2658 | Missense Mutation | CAG,CGG | Q,R 854 | NP_001265553.1 | |
NM_152995.5 | 2658 | Missense Mutation | CAG,CGG | Q,R 854 | NP_694540.3 |