Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCAGGGGAGGATTTGGACGAGGG[G/T]GTGGCCGCGGAGGCTTTAACAAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606468 | ||||||||||||||||||||
Literature Links: |
GAR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAR1 - GAR1 ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018983.3 | 464 | Missense Mutation | GGT,TGT | G,C 53 | NP_061856.1 | |
NM_032993.2 | 464 | Intron | NP_127460.1 |