Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGAGGAATACCGCCACATGCC[G/T]CTGCTGAAGCTCTGGGTCGGGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608614 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4V2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP4V2 - cytochrome P450 family 4 subfamily V member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207352.3 | 353 | Silent Mutation | CCG,CCT | P,P 88 | NP_997235.3 | |
XM_005262935.3 | 353 | Silent Mutation | CCG,CCT | P,P 88 | XP_005262992.1 | |
XM_017008037.1 | 353 | UTR 5 | XP_016863526.1 |
FLJ38576 - uncharacterized LOC651430 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |