Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGGAGGCCTTTCGGCTGGCGCA[C/G]CAGCCGCACCAGAACCAGGCGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606280 | ||||||||||||||||||||
Literature Links: |
DCAF16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAF16 - DDB1 and CUL4 associated factor 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017741.3 | 319 | Intron | NP_060211.3 | |||
XM_005248169.3 | 319 | Intron | XP_005248226.1 | |||
XM_005248171.3 | 319 | Intron | XP_005248228.1 | |||
XM_006713966.3 | 319 | Intron | XP_006714029.1 | |||
XM_017008327.1 | 319 | Intron | XP_016863816.1 | |||
XM_017008328.1 | 319 | Intron | XP_016863817.1 |
NCAPG - non-SMC condensin I complex subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022346.4 | 319 | Missense Mutation | CAC,CAG | H,Q 18 | NP_071741.2 | |
XM_011513876.2 | 319 | UTR 5 | XP_011512178.1 | |||
XM_017008543.1 | 319 | Missense Mutation | CAC,CAG | H,Q 18 | XP_016864032.1 | |
XM_017008544.1 | 319 | UTR 5 | XP_016864033.1 |