Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCGTTGGCGGAATTTGCAGTATT[G/T]CCTCATCATTCCGAAGGTGAGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607830 | ||||||||||||||||||||
Literature Links: |
FRAS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRAS1 - Fraser extracellular matrix complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166133.1 | 747 | Missense Mutation | TTG,TTT | L,F 20 | NP_001159605.1 | |
NM_025074.6 | 747 | Missense Mutation | TTG,TTT | L,F 20 | NP_079350.5 | |
XM_006714316.2 | 747 | Missense Mutation | TTG,TTT | L,F 20 | XP_006714379.1 |