Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610257 MIM: 612537 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC31A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC31A - SEC31 homolog A, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD143 - small nucleolar RNA, C/D box 143 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD144 - small nucleolar RNA, C/D box 144 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THAP9 - THAP domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317776.1 | 211 | UTR 5 | NP_001304705.1 | |||
NM_024672.5 | 211 | Silent Mutation | AAA,AAG | K,K 37 | NP_078948.3 | |
XM_005263220.2 | 211 | UTR 5 | XP_005263277.1 | |||
XM_011532246.2 | 211 | Intron | XP_011530548.1 | |||
XM_017008603.1 | 211 | Silent Mutation | AAA,AAG | K,K 50 | XP_016864092.1 | |
XM_017008604.1 | 211 | UTR 5 | XP_016864093.1 | |||
XM_017008605.1 | 211 | UTR 5 | XP_016864094.1 | |||
XM_017008606.1 | 211 | UTR 5 | XP_016864095.1 | |||
XM_017008607.1 | 211 | UTR 5 | XP_016864096.1 | |||
XM_017008608.1 | 211 | UTR 5 | XP_016864097.1 | |||
XM_017008609.1 | 211 | UTR 5 | XP_016864098.1 | |||
XM_017008610.1 | 211 | UTR 5 | XP_016864099.1 |
THAP9-AS1 - THAP9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |