Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCATAACTGCTATCAAACAAAT[A/C]ACCAGAATCTGTCCTCTGTCTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600345 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTC - betacellulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316963.1 | 705 | Intron | NP_001303892.1 | |||
NM_001729.3 | 705 | Silent Mutation | NP_001720.1 | |||
XM_011532211.1 | 705 | Silent Mutation | XP_011530513.1 | |||
XM_017008568.1 | 705 | Intron | XP_016864057.1 |