Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCATGCTGGACGACATGCTCAT[A/G]GACGAGATAGAGTTGGGTGGGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PITX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PITX2 - paired like homeodomain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000325.5 | 2108 | Silent Mutation | TCC,TCT | S,S 226 | NP_000316.2 | |
NM_001204397.1 | 2108 | Silent Mutation | TCC,TCT | S,S 219 | NP_001191326.1 | |
NM_001204398.1 | 2108 | Silent Mutation | TCC,TCT | S,S 219 | NP_001191327.1 | |
NM_001204399.1 | 2108 | Silent Mutation | TCC,TCT | S,S 173 | NP_001191328.1 | |
NM_153426.2 | 2108 | Silent Mutation | TCC,TCT | S,S 219 | NP_700475.1 | |
NM_153427.2 | 2108 | Silent Mutation | TCC,TCT | S,S 173 | NP_700476.1 | |
XM_011532027.2 | 2108 | Silent Mutation | TCC,TCT | S,S 173 | XP_011530329.1 |