Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAAATACTGATTCAAGTTGAAC[C/G]TCTGCCGGTTTCTTGATGAAGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAT1 - FAT atypical cadherin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005245.3 | 13636 | Missense Mutation | ACG,AGG | T,R 4483 | NP_005236.2 | |
XM_005262834.2 | 13636 | Missense Mutation | ACG,AGG | T,R 4495 | XP_005262891.1 | |
XM_005262835.2 | 13636 | Missense Mutation | ACG,AGG | T,R 4495 | XP_005262892.1 | |
XM_006714139.2 | 13636 | Missense Mutation | ACG,AGG | T,R 4483 | XP_006714202.1 |