Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATTTAATTCTTCATTGTATTC[C/T]GAATCCAGGGATGGGCTTGTGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102579 MIM: 608151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFC1 - replication factor C subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204747.1 | 3338 | Silent Mutation | TCA,TCG | S,S 1093 | NP_001191676.1 | |
NM_002913.4 | 3338 | Silent Mutation | TCA,TCG | S,S 1092 | NP_002904.3 | |
XM_011513730.1 | 3338 | Silent Mutation | TCA,TCG | S,S 1067 | XP_011512032.1 | |
XM_011513731.1 | 3338 | Silent Mutation | TCA,TCG | S,S 923 | XP_011512033.1 | |
XM_017008528.1 | 3338 | Silent Mutation | TCA,TCG | S,S 1066 | XP_016864017.1 |
WDR19 - WD repeat domain 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |