Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTGTAGATAGCCAAGTGTTTTC[A/G]TGGGGAAAGAACAGCCATGGGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HERC6 - HECT and RLD domain containing E3 ubiquitin protein ligase family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165136.1 | 833 | Silent Mutation | TCA,TCG | S,S 151 | NP_001158608.1 | |
NM_017912.3 | 833 | Silent Mutation | TCA,TCG | S,S 151 | NP_060382.3 | |
XM_005263083.3 | 833 | Silent Mutation | TCA,TCG | S,S 163 | XP_005263140.1 | |
XM_011532053.2 | 833 | Silent Mutation | TCA,TCG | S,S 163 | XP_011530355.1 | |
XM_017008333.1 | 833 | UTR 5 | XP_016863822.1 |