Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTCTCACTAAGTTTATGAATTA[A/G]AATGGCTAATTCCTCAGTTGCTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607590 MIM: 602345 | ||||||||||||||||||||
Literature Links: |
BBS7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BBS7 - Bardet-Biedl syndrome 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPC3 - transient receptor potential cation channel subfamily C member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130698.1 | 3141 | Silent Mutation | CTA,TTA | L,L 905 | NP_001124170.1 | |
NM_003305.2 | 3141 | Silent Mutation | CTA,TTA | L,L 832 | NP_003296.1 | |
XM_011532217.2 | 3141 | Silent Mutation | CTA,TTA | L,L 921 | XP_011530519.1 | |
XM_011532218.2 | 3141 | Silent Mutation | CTA,TTA | L,L 904 | XP_011530520.1 | |
XM_017008578.1 | 3141 | Silent Mutation | CTA,TTA | L,L 920 | XP_016864067.1 | |
XM_017008579.1 | 3141 | Silent Mutation | CTA,TTA | L,L 877 | XP_016864068.1 |