Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGCATGAAAAGTAGCGTCAAA[A/G]GAAAAGGACTCAAATTCTGTTGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP3 - caspase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004346.3 | 1016 | Silent Mutation | TCC,TCT | S,S 251 | NP_004337.2 | |
NM_032991.2 | 1016 | Silent Mutation | TCC,TCT | S,S 251 | NP_116786.1 | |
XM_011532301.1 | 1016 | Silent Mutation | TCC,TCT | S,S 251 | XP_011530603.1 |
LVCAT8 - liver cancer-associated transcript 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |