Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACACTTCCTGGAACTCCTCGGGC[A/G]ACAAGTGTCTCTTCAAACAAAAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612544 | ||||||||||||||||||||
Literature Links: |
ABLIM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABLIM2 - actin binding LIM protein family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130083.1 | 1605 | Missense Mutation | TCG,TTG | S,L 612 | NP_001123555.1 | |
NM_001130084.1 | 1605 | Missense Mutation | TCG,TTG | S,L 578 | NP_001123556.1 | |
NM_001130085.1 | 1605 | Missense Mutation | TCG,TTG | S,L 539 | NP_001123557.1 | |
NM_001130086.1 | 1605 | Missense Mutation | TCG,TTG | S,L 526 | NP_001123558.1 | |
NM_001130087.1 | 1605 | Missense Mutation | TCG,TTG | S,L 498 | NP_001123559.1 | |
NM_001130088.1 | 1605 | Intron | NP_001123560.1 | |||
NM_001286688.1 | 1605 | Missense Mutation | TCG,TTG | S,L 313 | NP_001273617.1 | |
NM_032432.4 | 1605 | Missense Mutation | TCG,TTG | S,L 488 | NP_115808.3 | |
XM_005248014.4 | 1605 | Missense Mutation | TCG,TTG | S,L 668 | XP_005248071.1 | |
XM_005248017.4 | 1605 | Missense Mutation | TCG,TTG | S,L 629 | XP_005248074.1 | |
XM_005248018.4 | 1605 | Missense Mutation | TCG,TTG | S,L 628 | XP_005248075.1 | |
XM_005248019.4 | 1605 | Missense Mutation | TCG,TTG | S,L 617 | XP_005248076.1 | |
XM_005248020.4 | 1605 | Missense Mutation | TCG,TTG | S,L 616 | XP_005248077.1 | |
XM_005248021.4 | 1605 | Missense Mutation | TCG,TTG | S,L 589 | XP_005248078.1 | |
XM_005248022.4 | 1605 | Missense Mutation | TCG,TTG | S,L 583 | XP_005248079.1 | |
XM_005248023.4 | 1605 | Missense Mutation | TCG,TTG | S,L 577 | XP_005248080.1 | |
XM_005248024.4 | 1605 | Missense Mutation | TCG,TTG | S,L 544 | XP_005248081.1 | |
XM_005248027.4 | 1605 | Missense Mutation | TCG,TTG | S,L 531 | XP_005248084.1 | |
XM_005248028.4 | 1605 | Missense Mutation | TCG,TTG | S,L 504 | XP_005248085.1 | |
XM_005248029.4 | 1605 | Missense Mutation | TCG,TTG | S,L 493 | XP_005248086.1 | |
XM_005248030.4 | 1605 | Missense Mutation | TCG,TTG | S,L 492 | XP_005248087.1 | |
XM_005248031.4 | 1605 | Intron | XP_005248088.1 | |||
XM_006713923.3 | 1605 | Missense Mutation | TCG,TTG | S,L 628 | XP_006713986.1 | |
XM_006713924.3 | 1605 | Missense Mutation | TCG,TTG | S,L 627 | XP_006713987.1 | |
XM_006713925.3 | 1605 | Missense Mutation | TCG,TTG | S,L 588 | XP_006713988.1 | |
XM_006713927.3 | 1605 | Missense Mutation | TCG,TTG | S,L 503 | XP_006713990.1 | |
XM_011513586.2 | 1605 | UTR 3 | XP_011511888.1 | |||
XM_017008706.1 | 1605 | Missense Mutation | TCG,TTG | S,L 667 | XP_016864195.1 | |
XM_017008707.1 | 1605 | Missense Mutation | TCG,TTG | S,L 624 | XP_016864196.1 | |
XM_017008708.1 | 1605 | Missense Mutation | TCG,TTG | S,L 618 | XP_016864197.1 | |
XM_017008709.1 | 1605 | Missense Mutation | TCG,TTG | S,L 595 | XP_016864198.1 | |
XM_017008710.1 | 1605 | Missense Mutation | TCG,TTG | S,L 594 | XP_016864199.1 | |
XM_017008711.1 | 1605 | Missense Mutation | TCG,TTG | S,L 585 | XP_016864200.1 | |
XM_017008712.1 | 1605 | Missense Mutation | TCG,TTG | S,L 584 | XP_016864201.1 | |
XM_017008713.1 | 1605 | Missense Mutation | TCG,TTG | S,L 578 | XP_016864202.1 | |
XM_017008714.1 | 1605 | Missense Mutation | TCG,TTG | S,L 573 | XP_016864203.1 | |
XM_017008715.1 | 1605 | Missense Mutation | TCG,TTG | S,L 559 | XP_016864204.1 | |
XM_017008716.1 | 1605 | Missense Mutation | TCG,TTG | S,L 556 | XP_016864205.1 | |
XM_017008717.1 | 1605 | Missense Mutation | TCG,TTG | S,L 555 | XP_016864206.1 | |
XM_017008718.1 | 1605 | Missense Mutation | TCG,TTG | S,L 550 | XP_016864207.1 | |
XM_017008719.1 | 1605 | Missense Mutation | TCG,TTG | S,L 545 | XP_016864208.1 | |
XM_017008720.1 | 1605 | Missense Mutation | TCG,TTG | S,L 542 | XP_016864209.1 | |
XM_017008721.1 | 1605 | Missense Mutation | TCG,TTG | S,L 532 | XP_016864210.1 | |
XM_017008722.1 | 1605 | Missense Mutation | TCG,TTG | S,L 526 | XP_016864211.1 | |
XM_017008723.1 | 1605 | Intron | XP_016864212.1 | |||
XM_017008724.1 | 1605 | Missense Mutation | TCG,TTG | S,L 499 | XP_016864213.1 | |
XM_017008725.1 | 1605 | Intron | XP_016864214.1 | |||
XM_017008726.1 | 1605 | Missense Mutation | TCG,TTG | S,L 379 | XP_016864215.1 | |
XM_017008727.1 | 1605 | Missense Mutation | TCG,TTG | S,L 340 | XP_016864216.1 |