Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGACATGTCCAGGTTAGAGCCT[A/T]TAACTGCAAAGCTTCCTGTCTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607520 | ||||||||||||||||||||
Literature Links: |
FRYL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRYL - FRY like transcription coactivator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015030.1 | 6193 | Missense Mutation | AAA,ATA | K,I 2966 | NP_055845.1 | |
XM_005248090.4 | 6193 | Missense Mutation | AAA,ATA | K,I 2981 | XP_005248147.1 | |
XM_005248093.4 | 6193 | Missense Mutation | AAA,ATA | K,I 1997 | XP_005248150.1 | |
XM_011513680.2 | 6193 | Missense Mutation | AAA,ATA | K,I 2980 | XP_011511982.1 | |
XM_011513681.2 | 6193 | Missense Mutation | AAA,ATA | K,I 2975 | XP_011511983.1 | |
XM_011513682.2 | 6193 | Missense Mutation | AAA,ATA | K,I 2960 | XP_011511984.1 | |
XM_011513683.2 | 6193 | Missense Mutation | AAA,ATA | K,I 2981 | XP_011511985.1 | |
XM_011513684.2 | 6193 | Missense Mutation | AAA,ATA | K,I 1997 | XP_011511986.1 | |
XM_011513685.2 | 6193 | Intron | XP_011511987.1 | |||
XM_017008046.1 | 6193 | Missense Mutation | AAA,ATA | K,I 2966 | XP_016863535.1 |
ZAR1 - zygote arrest 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175619.2 | 6193 | Intron | NP_783318.1 |