Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACCTGAATGGCAAGTTGAACCT[C/G]AGACTGCTTTCTGCTGTTTCTTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610010 | ||||||||||||||||||||
Literature Links: |
ARSJ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARSJ - arylsulfatase family member J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024590.3 | 3246 | Nonsense Mutation | TCA,TGA | S,* 588 | NP_078866.3 | |
XM_005263212.4 | 3246 | Nonsense Mutation | TCA,TGA | S,* 588 | XP_005263269.1 | |
XM_011532238.2 | 3246 | Nonsense Mutation | TCA,TGA | S,* 481 | XP_011530540.1 | |
XM_011532239.2 | 3246 | Nonsense Mutation | TCA,TGA | S,* 472 | XP_011530541.1 | |
XM_017008592.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 588 | XP_016864081.1 | |
XM_017008593.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 472 | XP_016864082.1 | |
XM_017008594.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 472 | XP_016864083.1 | |
XM_017008595.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 472 | XP_016864084.1 | |
XM_017008596.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 472 | XP_016864085.1 | |
XM_017008597.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 416 | XP_016864086.1 | |
XM_017008598.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 416 | XP_016864087.1 | |
XM_017008599.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 416 | XP_016864088.1 | |
XM_017008600.1 | 3246 | Nonsense Mutation | TCA,TGA | S,* 416 | XP_016864089.1 |